Anti-COL4A3BP Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-54HEA 3460 DKK
USBIC7510-54H
Anti-COL4A3BP Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:2000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: COL4A3BP (collagen, type IV, alpha 3 (Goodpasture antigen) binding protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR