Anti-COL4 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-52NEA 4320 DKK
USBIC7510-52N
Anti-COL4 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR