Anti-COL4A1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-52M-APCEA 6360 DKK
USBIC7510-52M-APC
Anti-COL4A1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antistoffer
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: COL4A1 (collagen, type IV, alpha 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR