Anti-COL4 Mouse Monoclonal Antibody (Biotin) [clone: 5K134]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-50R2EA 3310 DKK
USBIC7510-50R2
Anti-COL4 Mouse Monoclonal Antibody (Biotin) [clone: 5K134]
Antistoffer
Recommended dilution: ELISA: 1:2000-1:8000; Immunochemistry: ? 2?g/ml

Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: Monoclonal
Clone: 5K134
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR