Anti-COL3 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-44FEA 5280 DKK
USBIC7510-44F
Anti-COL3 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 2.5 ?g/ml; ELISA: 1:5000-1:10000; Western Blot: 1:5000-1:10000

Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR