Anti-COL2 Mouse Monoclonal Antibody [clone: 10B2285]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-20REA 5040 DKK
USBIC7510-20R
Anti-COL2 Mouse Monoclonal Antibody [clone: 10B2285]
Antistoffer
Recommended dilution: ELISA: 1:100-1:500; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: Monoclonal
Clone: 10B2285
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat, Sheep
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR