Anti-COL2 Mouse Monoclonal Antibody [clone: 9G171]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-20CEA 4410 DKK
USBIC7510-20C
Anti-COL2 Mouse Monoclonal Antibody [clone: 9G171]
Antistoffer
Recommended dilution: Immunochemistry: 1:10

Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: Monoclonal
Clone: 9G171
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Chicken, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR