Anti-COL2 Goat Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-19SEA 4070 DKK
USBIC7510-19S
Anti-COL2 Goat Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: Immunochemistry: 1:20-1:40; ELISA: 1:1000-1:4000

Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR