Anti-COL2 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-19REA 3620 DKK
USBIC7510-19R
Anti-COL2 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:10-1:20

Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR