Anti-COL2 Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-19L-50EA 2960 DKK
USBIC7510-19L-50 USBIC7510-19L-100
Anti-COL2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR