Anti-COL1 Mouse Monoclonal Antibody [clone: 9D7]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-14HEA 5380 DKK
USBIC7510-14H
Anti-COL1 Mouse Monoclonal Antibody [clone: 9D7]
Antistoffer
Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: Monoclonal
Clone: 9D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR