Anti-COL1 Mouse Monoclonal Antibody [clone: 10B2281]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7510-11GEA 4800 DKK
USBIC7510-11G
Anti-COL1 Mouse Monoclonal Antibody [clone: 10B2281]
Antistoffer
Recommended dilution: Immunochemistry: 10-20 ?g/ml

Type: Primary
Antigen: COL1 (collagen, type I)
Clonality: Monoclonal
Clone: 10B2281
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR