Anti-Cold Shock Domain Protein A Mouse monoclonal antibody [clone: 10B2613]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7508-18EA 0 DKK
USBIC7508-18
Anti-Cold Shock Domain Protein A Mouse monoclonal antibody [clone: 10B2613]
Type: Primary
Antigen: Cold Shock Domain Protein A
Clonality: Monoclonal
Clone: 10B2613
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR