Anti-CIRBP Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7508-01EA 4120 DKK
USBIC7508-01
Anti-CIRBP Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:6000; Western Blot: 0. 05-0.2?g/ml

Type: Primary
Antigen: CIRBP (cold inducible RNA binding protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR