Anti-COIL Mouse Monoclonal Antibody [clone: 13A1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7506-98JEA 3460 DKK
USBIC7506-98J
Anti-COIL Mouse Monoclonal Antibody [clone: 13A1]
Antistoffer
Recommended dilution: Western Blot: 1:2000

Type: Primary
Antigen: COIL (coilin)
Clonality: Monoclonal
Clone: 13A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR