Anti-COG7 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2280]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7506-77-HRPEA 0 DKK
USBIC7506-77-HRP
Anti-COG7 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2280]
Type: Primary
Antigen: COG7 (component of oligomeric golgi complex 7)
Clonality: Monoclonal
Clone: 10B2280
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR