Anti-CFL2 Goat polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7506-50FEA 4540 DKK
USBIC7506-50F
Anti-CFL2 Goat polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 1:128000; Western Blot: 0. 01-0. 3?g/ml

Type: Primary
Antigen: CFL2 (cofilin 2 (muscle))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR