Anti-CFL2 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2278]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7506-50E-APEA 0 DKK
USBIC7506-50E-AP
Anti-CFL2 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2278]
Type: Primary
Antigen: CFL2 (cofilin 2 (muscle))
Clonality: Monoclonal
Clone: 10B2278
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR