Anti-Cofilin 1/2 Rabbit Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC7506-49EA 2920 DKK
USBIC7506-49
Anti-Cofilin 1/2 Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: Western Blot: 1:500-1000; Immunochemistry: 1:50-1:100
Type: Primary
Antigen: Cofilin 1/2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human