Anti-CFL1 Mouse monoclonal antibody [clone: AT1C1]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7506-25FEA 4320 DKK
USBIC7506-25F
Anti-CFL1 Mouse monoclonal antibody [clone: AT1C1]
Antistoffer
Recommended dilution: Western Blot: 1:1000-1:2000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CFL1 (cofilin 1 (non-muscle))
Clonality: Monoclonal
Clone: AT1C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR