Anti-CFL1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7506-25BEA 2590 DKK
USBIC7506-25B
Anti-CFL1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: Immunochemistry:1:50-100

Type: Primary
Antigen: CFL1 (cofilin 1 (non-muscle))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR