Anti-CFL1 Rabbit polyclonal antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7506-10CEA 2560 DKK
USBIC7506-10C
Anti-CFL1 Rabbit polyclonal antibody
Antistoffer
Recommended dilution: ELISA: 0. 05-0.2?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: CFL1 (cofilin 1 (non-muscle))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR