Anti-Cofactor Required for Sp1 Transcriptional Activation Subunit 9 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2606]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7506-02A-APEA 0 DKK
USBIC7506-02A-AP
Anti-Cofactor Required for Sp1 Transcriptional Activation Subunit 9 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2606]
Type: Primary
Antigen: Cofactor Required for Sp1 Transcriptional Activation Subunit 9
Clonality: Monoclonal
Clone: 10B2606
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR