Anti-Cofactor Required for Sp1 Transcriptional Activation Subunit 6, 77kD Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2604]

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7506-01N-APEA 0 DKK
USBIC7506-01N-AP
Anti-Cofactor Required for Sp1 Transcriptional Activation Subunit 6, 77kD Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2604]
Type: Primary
Antigen: Cofactor Required for Sp1 Transcriptional Activation Subunit 6, 77kD
Clonality: Monoclonal
Clone: 10B2604
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR