Anti-Codeine Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7505-21AEA 6540 DKK
USBIC7505-21A
Anti-Codeine Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 0.5 ?g/ml

Type: Primary
Antigen: Codeine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR