Anti-COASY Mouse Monoclonal Antibody (Biotin) [clone: 10B2276]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC7480-BIOTINEA 4920 DKK
USBIC7480-BIOTIN
Anti-COASY Mouse Monoclonal Antibody (Biotin) [clone: 10B2276]
Antistoffer
Type: Primary
Antigen: COASY (CoA synthase)
Clonality: Monoclonal
Clone: 10B2276
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR