Anti-CNPase Chicken Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC6205-01AEA 2410 DKK
USBIC6205-01A
Anti-CNPase Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: CNPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR