Anti-CNPase Chicken Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC6200-01FEA 5940 DKK
USBIC6200-01F
Anti-CNPase Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 1:1000-1:2000; Western Blot: 1:2000-1:5000
Type: Primary
Antigen: CNPase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Mouse