Anti-CNBP Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5998-01CEA 3460 DKK
USBIC5998-01C
Anti-CNBP Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:8000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: CNBP (CCHC-type zinc finger, nucleic acid binding protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR