Anti-CMKLR1 Rat Monoclonal Antibody [clone: 10B2266]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5895-01H1EA 3160 DKK
USBIC5895-01H1
Anti-CMKLR1 Rat Monoclonal Antibody [clone: 10B2266]
Antistoffer
Recommended dilution: Flow Cytometry: 1:25-1:100

Type: Primary
Antigen: CMKLR1 (chemokine-like receptor 1)
Clonality: Monoclonal
Clone: 10B2266
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR