Anti-Cma2 Goat Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5886-20EA 3140 DKK
USBIC5886-20
Anti-Cma2 Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:2000

Type: Primary
Antigen: Cma2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR