Anti-CLP24 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 12K70]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5880-80EA 5030 DKK
USBIC5880-80
Anti-CLP24 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 12K70]
Antistoffer
Do not freeze!

Type: Primary
Antigen: CLP24
Clonality: Monoclonal
Clone: 12K70
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR