Anti-CLU Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5870-02EEA 5790 DKK
USBIC5870-02E
Anti-CLU Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: Immunochemistry: 5-15 ?g/ml

Type: Primary
Antigen: CLU (clusterin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR