Anti-CLU Goat Polyclonal Antibody
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC5870-01QEA 4730 DKK
USBIC5870-01Q
Anti-CLU Goat Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:32000; Western Blot: 1:32000; Immunochemistry: 2.5 ?g/ml
Type: Primary
Antigen: CLU (clusterin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human