Anti-CLU Mouse Monoclonal Antibody [clone: 10B2265]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5870-01GEA 6450 DKK
USBIC5870-01G
Anti-CLU Mouse Monoclonal Antibody [clone: 10B2265]
Antistoffer
Type: Primary
Antigen: CLU (clusterin)
Clonality: Monoclonal
Clone: 10B2265
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR