Anti-CLU Mouse Monoclonal Antibody [clone: 5i231]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5870-01EA 3570 DKK
USBIC5870-01
Anti-CLU Mouse Monoclonal Antibody [clone: 5i231]
Antistoffer
Type: Primary
Antigen: CLU (clusterin)
Clonality: Monoclonal
Clone: 5i231
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR