Anti-Clostridium difficile Toxin B Mouse Monoclonal Antibody [clone: 9L115]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5856-01PEA 4380 DKK
USBIC5856-01P
Anti-Clostridium difficile Toxin B Mouse Monoclonal Antibody [clone: 9L115]
Antistoffer
Type: Primary
Antigen: Clostridium difficile Toxin B
Clonality: Monoclonal
Clone: 9L115
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR