Anti-Clostridium difficile Toxin A Mouse Monoclonal Antibody [clone: 8J1029]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5856-01AEA 5530 DKK
USBIC5856-01A
Anti-Clostridium difficile Toxin A Mouse Monoclonal Antibody [clone: 8J1029]
Antistoffer
Type: Primary
Antigen: Clostridium difficile Toxin A
Clonality: Monoclonal
Clone: 8J1029
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR