Anti-Clostridium sp. Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5853-25EEA 3650 DKK
USBIC5853-25E
Anti-Clostridium sp. Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Clostridium sp.
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR