Anti-Clostridium perfringens Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5852-60EA 4380 DKK
USBIC5852-60
Anti-Clostridium perfringens Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:5000-1:20000

Type: Primary
Antigen: Clostridium perfringens
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR