Anti-Clostridium difficile Toxin A Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5851-95HEA 3190 DKK
USBIC5851-95H
Anti-Clostridium difficile Toxin A Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Clostridium difficile Toxin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR