Anti-Clostridium botulinum E Toxoid Mouse Monoclonal Antibody [clone: 7C223]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5851-82HEA 3700 DKK
USBIC5851-82H
Anti-Clostridium botulinum E Toxoid Mouse Monoclonal Antibody [clone: 7C223]
Antistoffer
Recommended dilution: Immunofluorescence: 1:25

Type: Primary
Antigen: Clostridium botulinum E Toxoid
Clonality: Monoclonal
Clone: 7C223
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR