Anti-Clostridium botulinum E Toxoid Mouse Monoclonal Antibody [clone: 7C222]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC5851-82GEA 3700 DKK
USBIC5851-82G
Anti-Clostridium botulinum E Toxoid Mouse Monoclonal Antibody [clone: 7C222]
Antistoffer
Recommended dilution: Immunofluorescence: 1:50
Type: Primary
Antigen: Clostridium botulinum E Toxoid
Clonality: Monoclonal
Clone: 7C222
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestil nu