Anti-Clostridium botulinum E Toxoid Mouse Monoclonal Antibody [clone: 11D4]

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5851-82FEA 5010 DKK
USBIC5851-82F
Anti-Clostridium botulinum E Toxoid Mouse Monoclonal Antibody [clone: 11D4]
Antistoffer
  • Recommended dilution: ELISA: 1:4000

Type: Primary
Antigen: Clostridium botulinum E Toxoid
Clonality: Monoclonal
Clone: 11D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR