Anti-Clostridium botulinum Toxin A Rabbit Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5851-80FEA 3230 DKK
USBIC5851-80F
Anti-Clostridium botulinum Toxin A Rabbit Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:10000

Type: Primary
Antigen: Clostridium botulinum Toxin A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR