Anti-Clostridium botulinum Neurotoxin Type C1 Heavy Chain Mouse Monoclonal Antibody [clone: 11n49]
Leverandør: United States Biological
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
USBIC5851-49JEA 5440 DKK
USBIC5851-49J
Anti-Clostridium botulinum Neurotoxin Type C1 Heavy Chain Mouse Monoclonal Antibody [clone: 11n49]
Antistoffer
Recommended dilution: Western Blot: 1 ?g/ml; Immunoprecipitation: 25 ?g/ml
Type: Primary
Antigen: Clostridium botulinum Neurotoxin Type C1 Heavy Chain
Clonality: Monoclonal
Clone: 11n49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestil nu