Anti-Clostridium Neurotoxin Type C1 Heavy Chain Sheep Polyclonal Antibody (Biotin)

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5851-49EEA 5230 DKK
USBIC5851-49E
Anti-Clostridium Neurotoxin Type C1 Heavy Chain Sheep Polyclonal Antibody (Biotin)
Antistoffer
Recommended dilution: Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Clostridium Neurotoxin Type C1 Heavy Chain
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: E. coli
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR