Anti-Clostridium botulinum Neurotoxin Type C1 Heavy Chain Sheep Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5851-49BEA 5640 DKK
USBIC5851-49B
Anti-Clostridium botulinum Neurotoxin Type C1 Heavy Chain Sheep Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Clostridium botulinum Neurotoxin Type C1 Heavy Chain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR