Anti-Clostridium botulinum Neurotoxin Type A Heavy Chain Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5851-48HEA 3940 DKK
USBIC5851-48H
Anti-Clostridium botulinum Neurotoxin Type A Heavy Chain Chicken Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Primary
Antigen: Clostridium botulinum Neurotoxin Type A Heavy Chain
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR