Anti-Clostridium botulinum Neurotoxin Type A Heavy Chain Chicken Polyclonal Antibody

Leverandør: United States Biological
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

USBIC5851-48FEA 3680 DKK
USBIC5851-48F
Anti-Clostridium botulinum Neurotoxin Type A Heavy Chain Chicken Polyclonal Antibody
Antistoffer
Recommended dilution: ELISA: 1:100- 1:200; ELISA: 1:50000- 1:75000

Type: Primary
Antigen: Clostridium botulinum Neurotoxin Type A Heavy Chain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR